Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sophoricoside 99+%

Supplier: AMBEED, INC
Warning

A257552-100G A257552-25G A257552-500G A257552-50MG A257552-5G A257552-10MG
77289-144EA 233 USD InStock
77289-144 77289-146 77289-148 77289-150 77289-152 77748-482
Sophoricoside 99+%
Sophoricoside
Precaución: for Research Use Only


Formula: C₂₁H₂₀O₁₀
MDL Number: MFCD01075138
CAS Number: 152-95-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty