Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-chloro-1H-indole-2-carboxylate 98%

Supplier: AMBEED, INC
Warning

A258014-100G A258014-10G A258014-25G A258014-5G
77276-456EA 113 USD InStock
77276-456 77276-458 77276-460 77276-462
Ethyl-5-chloro-1H-indole-2-carboxylate 98%
Ethyl-5-chloro-1H-indole-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₀ClNO₂
MDL Number: MFCD00005610
CAS Number: 4792-67-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty