Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloronaphthalen-1-amine 97%

Supplier: AMBEED, INC
Warning

A258153-1G A258153-250MG A258153-25G A258153-5G A258153-10G
77262-134EA 20 USD InStock
77262-134 77262-136 77262-138 77262-140 77748-485
2,4-Dichloronaphthalen-1-amine 97%
2,4-Dichloronaphthalen-1-amine
Precaución: for Research Use Only


Formula: C₁₀H₇Cl₂N
MDL Number: MFCD19302400
CAS Number: 29242-87-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty