Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxybenzenesulphonyl chloride 97%

Supplier: AMBEED, INC
Danger

A258727-1G A258727-250MG A258727-25G A258727-5G
77276-492EA 22 USD InStock
77276-492 77276-494 77276-496 77276-498
3-Methoxybenzenesulphonyl chloride 97%
3-Methoxybenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₇H₇ClO₃S
MDL Number: MFCD01318155
CAS Number: 10130-74-2
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty