Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethoxybenzonitrile 98%

Supplier: AMBEED, INC
Warning

A259206-10G A259206-1G A259206-25G A259206-5G
77276-512EA 28 USD InStock
77276-512 77276-514 77276-516 77276-518
2,4-Dimethoxybenzonitrile 98%
2,4-Dimethoxybenzonitrile
Precaución: for Research Use Only


Formula: C₉H₉NO₂
MW: 163.18
Boiling Pt: 283
Melting Pt: 96.5
MDL Number: MFCD00001786
CAS Number: 4107-65-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty