Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-nitrophenol 97%

Supplier: AMBEED, INC
Danger

A259356-100G A259356-10G A259356-1G A259356-25G A259356-500G
77289-170EA 154 USD InStock
77289-170 77289-172 77289-174 77289-176 77289-178
3-Fluoro-4-nitrophenol 97%
3-Fluoro-4-nitrophenol
Precaución: for Research Use Only


Formula: C₆H₄FNO₃
MW: 157.1 g/mol
MDL Number: MFCD00041251
CAS Number: 394-41-2
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty