Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

VX-11e 98%

Supplier: AMBEED, INC
Warning

A260385-10MG A260385-1MG A260385-5MG
77238-488EA 136 USD InStock
77238-488 77238-490 77238-492
VX-11e 98%
VX-11e
Precaución: for Research Use Only


Formula: C₂₄H₂₀Cl₂FN₅O₂
MDL Number: MFCD18433366
CAS Number: 896720-20-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty