Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-4'-hexylbiphenyl 98% (by GC)

Supplier: AMBEED, INC
Warning

A260437-100MG A260437-1G A260437-250MG A260437-5G
77289-198EA 19 USD InStock
77289-198 77289-200 77289-202 77289-204
4-Cyano-4'-hexylbiphenyl 98% (by GC)
4-Cyano-4'-hexylbiphenyl
Precaución: for Research Use Only


Formula: C₁₉H₂₁N
MDL Number: MFCD00075144
CAS Number: 41122-70-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty