Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone 98%

Supplier: AMBEED, INC
Danger

A261200-1G A261200-250MG A261200-5G A261200-25G
77262-220EA 52 USD InStock
77262-220 77262-222 77262-224 77746-431
2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone 98%
2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone
Precaución: for Research Use Only


Formula: C₁₆H₂₃BrO₂
MDL Number: MFCD00173889
CAS Number: 14386-64-2
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty