Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Propanesulphonyl chloride 97%

Supplier: AMBEED, INC
Danger

A261628-10G A261628-25G A261628-5G A261628-100G
77238-504EA 10 USD InStock
77238-504 77238-506 77238-508 77746-433
1-Propanesulphonyl chloride 97%
1-Propanesulphonyl chloride
Precaución: for Research Use Only


Formula: C₃H₇ClO₂S
MW: 142.61 g/mol
Boiling Pt: 192
Melting Pt: -46
Flash Pt: 86
MDL Number: MFCD00007462
CAS Number: 10147-36-1
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty