Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(p-Toluenesulfonyl)methyl isocyanide 98%

Supplier: AMBEED, INC
Danger

A261697-10G A261697-5G A261697-25G
77276-554EA 15 USD InStock
77276-554 77276-556 77304-818
(p-Toluenesulfonyl)methyl isocyanide 98%
(p-Toluenesulphonyl)methyl isocyanide
Precaución: for Research Use Only


Formula: C₉H₉NO₂S
MDL Number: MFCD00000005
CAS Number: 36635-61-7
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty