Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MK-8931 98%

Supplier: AMBEED, INC
Warning

A262054-10MG A262054-1MG A262054-25MG A262054-50MG A262054-5MG
77250-168EA 162 USD InStock
77250-168 77250-170 77250-172 77250-174 77250-176
MK-8931 98%
MK-8931
Precaución: for Research Use Only


Formula: C₁₇H₁₇F₂N₅O₃S
MW: 409.41 g/mol
MDL Number: MFCD28963974
CAS Number: 1286770-55-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty