Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Chloro-5-fluorophenyl)acetic acid 95%

Supplier: AMBEED, INC
Warning

A262260-100G A262260-10G A262260-1G A262260-250MG A262260-25G A262260-5G
77276-582EA 640 USD InStock
77276-582 77276-584 77276-586 77276-588 77276-590 77276-592
(3-Chloro-5-fluorophenyl)acetic acid 95%
(3-Chloro-5-fluorophenyl)acetic acid
Precaución: for Research Use Only


Formula: (C₆H₃ClF)CH₂COOH
MW: 188.59
MDL Number: MFCD03788475
CAS Number: 202001-00-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty