Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(4-(tert-butyl)phenoxy)acetate 97%

Supplier: AMBEED, INC
Warning

A262654-100MG A262654-1G A262654-250MG
77262-254EA 5 USD InStock
77262-254 77262-256 77262-258
Methyl 2-(4-(tert-butyl)phenoxy)acetate 97%
Methyl 2-(4-(tert-butyl)phenoxy)acetate
Precaución: for Research Use Only


Formula: C₁₃H₁₈O₃
MDL Number: MFCD09909448
CAS Number: 88530-52-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty