Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CGM097 ≥98%

Supplier: AMBEED, INC
Warning

A263431-10MG A263431-1MG A263431-25MG A263431-2MG A263431-50MG A263431-5MG
77238-568EA 242.01 USD InStock
77238-568 77238-570 77238-572 77238-574 77238-576 77238-578
CGM097 ≥98%
CGM097
Precaución: for Research Use Only


Formula: C₃₈H₄₇ClN₄O₄
MDL Number: MFCD28144684
CAS Number: 1313363-54-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty