Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-fluorocinnoline 97%

Supplier: AMBEED, INC
Danger

A264042-100MG A264042-1G A264042-250MG
77262-300EA 59 USD InStock
77262-300 77262-302 77262-304
4-Chloro-6-fluorocinnoline 97%
4-Chloro-6-fluorocinnoline
Precaución: for Research Use Only


Formula: C₈H₄ClFN₂
MW: 182.58 g/mol
MDL Number: MFCD28040483
CAS Number: 2251-62-9
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty