Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-3-(2-Chloro-4-fluorophenyl)acrylic acid 98%

Supplier: AMBEED, INC
Warning

A264865-100MG A264865-1G A264865-25G A264865-5G
77262-320EA 5 USD InStock
77262-320 77262-322 77262-324 77262-326
(E)-3-(2-Chloro-4-fluorophenyl)acrylic acid 98%
(E)-3-(2-Chloro-4-fluorophenyl)acrylic acid
Precaución: for Research Use Only


Formula: C₉H₆ClFO₂
MDL Number: MFCD00236027
CAS Number: 174603-37-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty