Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,4-Trifluoro-1-butanol 98%

Supplier: AMBEED, INC
Danger

A264915-10G A264915-1G A264915-25G A264915-5G
77289-298EA 23 USD InStock
77289-298 77289-300 77289-302 77289-304
4,4,4-Trifluoro-1-butanol 98%
4,4,4-Trifluoro-1-butanol
Precaución: for Research Use Only


Formula: C₄H₇F₃O
MDL Number: MFCD00041483
CAS Number: 461-18-7
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty