Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(p-Toluenesulfonyl)ethanol 98%

Supplier: AMBEED, INC
Warning

A265149-100MG A265149-10G A265149-1G A265149-250MG A265149-5G
77250-232EA 5 USD InStock
77250-232 77250-234 77250-236 77250-238 77250-240
2-(p-Toluenesulfonyl)ethanol 98%
2-(p-Toluenesulphonyl)ethanol
Precaución: for Research Use Only


Formula: C₉H₁₂O₃S
MDL Number: MFCD00041757
CAS Number: 22381-54-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty