Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate 97%

Supplier: AMBEED, INC
Warning

A265302-10G A265302-25G A265302-5G
77289-318EA 19 USD InStock
77289-318 77289-320 77289-322
4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate 97%
4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate
Precaución: for Research Use Only


Formula: C₃₁H₄₈O₂
MW: 452.723 g/mol
MDL Number: MFCD00673705
CAS Number: 88038-92-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty