Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-N-(3-(2-chlorobenzoyl)-5-ethylthiophen-2-yl)acetamide 97%

Supplier: AMBEED, INC
Warning

A265511-1G A265511-250MG A265511-5G
77250-256EA 16 USD InStock
77250-256 77250-258 77250-260
2-Chloro-N-(3-(2-chlorobenzoyl)-5-ethylthiophen-2-yl)acetamide 97%
2-Chloro-N-(3-(2-chlorobenzoyl)-5-ethylthiophen-2-yl)acetamide
Precaución: for Research Use Only


Formula: C₁₅H₁₃Cl₂NO₂S
MDL Number: MFCD18969310
CAS Number: 50508-89-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty