Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GSK-269962 98%

Supplier: AMBEED, INC
Warning

A266066-1MG A266066-5MG
77238-634EA 48 USD InStock
77238-634 77238-636
GSK-269962 98%
GSK-269962
Precaución: for Research Use Only


Formula: C₂₉H₃₀N₈O₅
MDL Number: MFCD09970310
CAS Number: 850664-21-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty