Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorobenzenesulphonyl chloride 96%

Supplier: AMBEED, INC
Danger

A266144-10G A266144-25G A266144-5G
77289-356EA 10 USD InStock
77289-356 77289-358 77289-360
2-Fluorobenzenesulphonyl chloride 96%
2-Fluorobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₆H₄ClFO₂S
MW: 194.61
Boiling Pt: 242
Density: 1.47
MDL Number: MFCD00042285
CAS Number: 2905-21-7
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty