Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-1,2,3-Benzotriazole-5-carbonitrile 98%

Supplier: AMBEED, INC
Warning

A267515-250MG A267515-1G
77238-660EA 15 USD InStock
77238-660 77746-463
1H-1,2,3-Benzotriazole-5-carbonitrile 98%
1H-1,2,3-Benzotriazole-5-carbonitrile
Precaución: for Research Use Only


Formula: C₇H₄N₄
MDL Number: MFCD08669484
CAS Number: 24611-70-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty