Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Butyl-2-pyrrolidone 99%

Supplier: AMBEED, INC
Warning

A268002-100G A268002-25G A268002-500G A268002-5G
77238-662EA 60 USD InStock
77238-662 77238-664 77238-666 77238-668
1-Butyl-2-pyrrolidone 99%
1-Butyl-2-pyrrolidone
Precaución: for Research Use Only


Formula: C₈H₁₅NO
MDL Number: MFCD00020876
CAS Number: 3470-98-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty