Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate 98%

Supplier: AMBEED, INC
Warning

A268640-100MG A268640-1G A268640-250MG
77250-290EA 101 USD InStock
77250-290 77250-292 77250-294
(S)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate 98%
(S)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate
Precaución: for Research Use Only


Formula: C₃₆H₂₅F₁₂O₄P
MW: 780.53 g/mol
CAS Number: 2229836-07-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty