Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[(3-Triethoxysilyl)propyl]succinic anhydride 98%

Supplier: AMBEED, INC
Warning

A268822-1G A268822-25G A268822-5G
77276-726EA 13 USD InStock
77276-726 77276-728 77276-730
[(3-Triethoxysilyl)propyl]succinic anhydride 98%
[(3-Triethoxysilyl)propyl]succinic anhydride
Precaución: for Research Use Only


Formula: C₁₃H₂₄O₆Si
MW: 304.41 g/mol
MDL Number: MFCD01320976
CAS Number: 93642-68-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty