Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethoxyphenyl isothiocyanate 98%

Supplier: AMBEED, INC
Danger

A269770-1G A269770-25G A269770-5G
77276-738EA 11 USD InStock
77276-738 77276-740 77276-742
2,4-Dimethoxyphenyl isothiocyanate 98%
2,4-Dimethoxyphenyl isothiocyanate
Precaución: for Research Use Only


Formula: C₉H₉NO₂S
MW: 195.24 g/mol
Melting Pt: 57
MDL Number: MFCD00041058
CAS Number: 33904-03-9
UN: 1759
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty