Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-N-(o-tolyl)benzamide 97%

Supplier: AMBEED, INC
Warning

A269783-1G A269783-25G A269783-5G
77250-326EA 12 USD InStock
77250-326 77746-470 77746-471
2-Methyl-N-(o-tolyl)benzamide 97%
2-Methyl-N-(o-tolyl)benzamide
Precaución: for Research Use Only


Formula: C₁₅H₁₅NO
MW: 225.29 g/mol
MDL Number: MFCD00442534
CAS Number: 22978-49-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty