Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Aminopropyl)piperidine 97%

Supplier: AMBEED, INC
Danger

A270312-1G A270312-250MG A270312-25G A270312-5G
77289-434EA 33 USD InStock
77289-434 77289-436 77289-438 77289-440
1-(3-Aminopropyl)piperidine 97%
1-(3-Aminopropyl)piperidine
Precaución: for Research Use Only


Formula: C₈H₁₈N₂
MDL Number: MFCD00023784
CAS Number: 3529-08-6
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty