Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitro-1,3,5-trimethoxybenzene 98%

Supplier: AMBEED, INC
Warning

A271300-100G A271300-1G A271300-25G A271300-5G
77250-352EA 620 USD InStock
77250-352 77250-354 77250-356 77250-358
2-Nitro-1,3,5-trimethoxybenzene 98%
2-Nitro-1,3,5-trimethoxybenzene
Precaución: for Research Use Only


Formula: C₉H₁₁NO₅
MDL Number: MFCD00016992
CAS Number: 14227-18-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty