Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-methoxypyrazine 97%

Supplier: AMBEED, INC
Warning

A271798-10G A271798-1G A271798-250MG A271798-25G A271798-5G
77262-470EA 151 USD InStock
77262-470 77262-472 77262-474 77262-476 77262-478
2-Bromo-5-methoxypyrazine 97%
2-Bromo-5-methoxypyrazine
Precaución: for Research Use Only


Formula: C₅H₅BrN₂O
MDL Number: MFCD08235189
CAS Number: 143250-10-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty