Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodo-1-methoxy-3-nitrobenzene 97%

Supplier: AMBEED, INC
Warning

A272369-1G A272369-250MG A272369-5G
77262-480EA 12 USD InStock
77262-480 77262-482 77262-484
2-Iodo-1-methoxy-3-nitrobenzene 97%
2-Iodo-1-methoxy-3-nitrobenzene
Precaución: for Research Use Only


Formula: C₇H₆INO₃
MW: 279.03 g/mol
MDL Number: MFCD11656709
CAS Number: 98991-08-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty