Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloropicolinic acid 97%

Supplier: AMBEED, INC
Warning

A272399-100G A272399-1G A272399-25G A272399-500G A272399-5G A272399-10G
77276-792EA 74 USD InStock
77276-792 77276-794 77276-796 77276-798 77276-800 77748-522
6-Chloropicolinic acid 97%
6-Chloropicolinic acid
Precaución: for Research Use Only


Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
MDL Number: MFCD00155390
CAS Number: 4684-94-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty