Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methoxy-5-methylphenazinium methyl sulphate 96%

Supplier: AMBEED, INC
Warning

A272725-100MG A272725-1MG A272725-1G A272725-250MG
77238-780EA 112 USD InStock
77238-780 77238-782 77746-482 77746-483
1-Methoxy-5-methylphenazinium methyl sulphate 96%
1-Methoxy-5-methylphenazinium methyl sulphate
Precaución: for Research Use Only


Formula: C₁₅H₁₆N₂O₅S
MW: 336.37 g/mol
MDL Number: MFCD00040648
CAS Number: 65162-13-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty