Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorobenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A273523-100G A273523-10G A273523-25G A273523-500G
77262-528EA 27 USD InStock
77262-528 77262-530 77262-532 77262-534
2-Chlorobenzyl alcohol 98%
2-Chlorobenzyl alcohol


Formula: C₇H₇ClO
MDL Number: MFCD00004604
CAS Number: 17849-38-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty