Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-4-[2-(Diphenylphosphino)-1-naphthalenyl]-N-[(R)-1-phenylethoxy]phthalazine 97%

Supplier: AMBEED, INC
Warning

A276610-100MG A276610-1G A276610-250MG A276610-50MG
77250-426EA 83 USD InStock
77250-426 77250-428 77250-430 77250-432
(S)-4-[2-(Diphenylphosphino)-1-naphthalenyl]-N-[(R)-1-phenylethoxy]phthalazine 97%
(S)-4-[2-(Diphenylphosphino)-1-naphthalenyl]-N-[(R)-1-phenylethoxy]phthalazine
Precaución: for Research Use Only


Formula: C₃₈H₂₉N₂OP
CAS Number: 828927-94-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty