Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-(((6R,7R)-7-Amino-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)thio)thiazol-4-yl)-1-methylpyridin-1-ium chloride hydrochloride 97%

Supplier: AMBEED, INC
Warning

A276779-10G A276779-1G A276779-5G
77250-446EA 116 USD InStock
77250-446 77250-448 77250-450 77250-452
4-(2-(((6R,7R)-7-Amino-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)thio)thiazol-4-yl)-1-methylpyridin-1-ium chloride hydrochloride 97%
4-(2-(((6R,7R)-7-Amino-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)thio)thiazol-4-yl)-1-methylpyridin-1-ium chloride hydrochloride
Precaución: for Research Use Only


Formula: C₁₆H₁₆Cl₂N₄O₃S₃
CAS Number: 400827-64-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty