Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-4'-pentylbiphenyl 98%

Supplier: AMBEED, INC
Warning

A278035-100G A278035-10G A278035-250MG A278035-25G A278035-5G A278035-1G
77289-576EA 57 USD InStock
77289-576 77289-578 77289-580 77289-582 77289-584 77748-534
4-Bromo-4'-pentylbiphenyl 98%
4-Bromo-4'-pentylbiphenyl
Precaución: for Research Use Only


Formula: C₁₇H₁₉Br
MW: 303.2400 g/mol
MDL Number: MFCD00060106
CAS Number: 63619-59-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty