Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromopropene 90%

Supplier: AMBEED, INC
Danger

A278259-25G A278259-5G
77250-472EA 13 USD InStock
77250-472 77250-474
2,3-Dibromopropene 90%
2,3-Dibromopropene
Precaución: for Research Use Only


Formula: C₃H₄Br₂
MDL Number: MFCD00000211
CAS Number: 513-31-5
UN: 3334
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty