Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1E,6E)-1,7-Bis(3,4-dimethoxyphenyl)hepta-1,6-diene-3,5-dione 98%

Supplier: AMBEED, INC
Warning

A279009-10MG A279009-1MG A279009-5MG
77262-606EA 161 USD InStock
77262-606 77262-608 77262-610
(1E,6E)-1,7-Bis(3,4-dimethoxyphenyl)hepta-1,6-diene-3,5-dione 98%
(1E,6E)-1,7-Bis(3,4-dimethoxyphenyl)hepta-1,6-diene-3,5-dione
Precaución: for Research Use Only


Formula: C₂₃H₂₄O₆
MDL Number: MFCD28964095
CAS Number: 160096-59-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty