Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one 98+%

Supplier: AMBEED, INC
Warning

A279135-100MG A279135-10MG A279135-1MG A279135-25MG A279135-50MG A279135-5MG
77238-846EA 58 USD InStock
77238-846 77238-848 77238-850 77238-852 77238-854 77238-856
2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one 98+%
2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one
Precaución: for Research Use Only


Formula: C₁₆H₁₅N₃O₂
MDL Number: MFCD10568147
CAS Number: 168425-64-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty