Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Citraconic anhydride 97%

Supplier: AMBEED, INC
Danger

A281350-100G A281350-10G
77276-970EA 36 USD InStock
77276-970 77276-972 77276-974
Citraconic anhydride 97%
Citraconic anhydride
Precaución: for Research Use Only


Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 213
Flash Pt: 101
MDL Number: MFCD00005522
CAS Number: 616-02-4
UN: 2922
ADR: 8,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty