Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzoyl benzylamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A282045-100MG A282045-1G A282045-250MG
77238-892EA 91 USD InStock
77238-892 77238-894 77238-896
4-Benzoyl benzylamine hydrochloride 95%
4-Benzoyl benzylamine hydrochloride
Precaución: for Research Use Only


Formula: C₁₄H₁₄ClNO
MW: 247.73 g/mol
MDL Number: MFCD06245504
CAS Number: 24095-40-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty