Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-2-(1H)-pyridone 98%

Supplier: AMBEED, INC
Warning

A282479-1G A282479-250MG A282479-25G A282479-5G
77289-676EA 10 USD InStock
77289-676 77289-678 77289-680 77289-682
4-Benzyloxy-2-(1H)-pyridone 98%
4-Benzyloxy-2-(1H)-pyridone
Precaución: for Research Use Only


Formula: C₁₂H₁₁NO₂
MW: 201.22 g/mol
MDL Number: MFCD04039771
CAS Number: 53937-02-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty