Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxypentanoic acid 97%

Supplier: AMBEED, INC
Warning

A282505-1G A282505-250MG A282505-5G
77276-984EA 270 USD InStock
77276-984 77276-986 77748-550
3-Hydroxypentanoic acid 97%
3-Hydroxypentanoic acid
Precaución: for Research Use Only


Formula: C₅H₁₀O₃
MW: 118.13 g/mol
MDL Number: MFCD07778468
CAS Number: 10237-77-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty