Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydro-1,1-dioxo-1,2-benzisothiazole 96%

Supplier: AMBEED, INC
Warning

A282871-100MG A282871-1G A282871-250MG A282871-5G
77250-536EA 27 USD InStock
77250-536 77250-538 77250-540 77250-542
2,3-Dihydro-1,1-dioxo-1,2-benzisothiazole 96%
2,3-Dihydro-1,1-dioxo-1,2-benzisothiazole
Precaución: for Research Use Only


Formula: C₇H₇NO₂S
MW: 169.201 g/mol
MDL Number: MFCD03426201
CAS Number: 936-16-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty