Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-isopropylaniline 97%

Supplier: AMBEED, INC
Warning

A283201-100MG A283201-250MG A283201-25MG A283201-50MG
77238-908EA 14 USD InStock
77238-908 77238-910 77238-912 77238-914
4-Fluoro-2-isopropylaniline 97%
4-Fluoro-2-isopropylaniline
Precaución: for Research Use Only


Formula: C₉H₁₂FN
MW: 153.19 g/mol
MDL Number: MFCD19598718
CAS Number: 1339874-93-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty