Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxyflavone 95%

Supplier: AMBEED, INC
Warning

A284723-10G A284723-1G A284723-250MG A284723-5G
77277-038EA 75 USD InStock
77277-038 77277-040 77277-042 77277-044
3-Hydroxyflavone 95%
3-Hydroxyflavone
Precaución: for Research Use Only


Formula: C₁₅H₁₀O₃
MW: 238.24 g/mol
Boiling Pt: 411
MDL Number: MFCD00006832
CAS Number: 577-85-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty