Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Dimethylamino)-4,6-pyrimidinediol 95%

Supplier: AMBEED, INC
Warning

A284920-100MG A284920-250MG
77250-570EA 48 USD InStock
77250-570 77250-572
2-(Dimethylamino)-4,6-pyrimidinediol 95%
2-(Dimethylamino)-4,6-pyrimidinediol
Precaución: for Research Use Only


Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
MDL Number: MFCD01935970
CAS Number: 5738-14-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty